Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGGGCTCTGCTACTTACAGATG[C/T]TCACATGGCTCTGCAGCTCTCCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611158 | ||||||||||||||||||||
Literature Links: |
KRT77 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRT77 - keratin 77 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175078.2 | 1248 | Missense Mutation | AAC,AGC | N,S 486 | NP_778253.2 | |
XM_011538288.2 | 1248 | Missense Mutation | AAC,AGC | N,S 253 | XP_011536590.1 | |
XM_011538289.2 | 1248 | Intron | XP_011536591.1 |