Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGATGCCACCTCCAAAGCTGGC[C/T]GCGCCACCTCCGCACACAGTGGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123940 MIM: 611160 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRT4 - keratin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRT79 - keratin 79 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175834.2 | 1476 | Silent Mutation | GCA,GCG | A,A 481 | NP_787028.1 |