Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCAGGTAACCCAAGTGAATTTGC[C/T]ATCTCTGAGGAAGTCATGGCCTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603425 MIM: 612822 | ||||||||||||||||||||
Literature Links: |
ARL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARL1 - ADP ribosylation factor like GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177.5 | 592 | Missense Mutation | ATA,ATG | I,M 139 | NP_001168.1 | |
NM_001301068.1 | 592 | Missense Mutation | ATA,ATG | I,M 93 | NP_001287997.1 |
UTP20 - UTP20, small subunit processome component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |