Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACAGAGGGTGGCCGGTCCCCAGA[C/T]GCCAGGCCCGGGGCCGGGGACTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600848 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCOR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCOR2 - nuclear receptor corepressor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077261.3 | 7410 | Silent Mutation | GCA,GCG | A,A 2363 | NP_001070729.2 | |
NM_001206654.1 | 7410 | Silent Mutation | GCA,GCG | A,A 2409 | NP_001193583.1 | |
NM_006312.5 | 7410 | Silent Mutation | GCA,GCG | A,A 2419 | NP_006303.4 |