Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCGCGGAGCACGCGTCCCAGCAG[C/T]TCCAGCGTGGCCTCGCGCCGCGGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611743 MIM: 191190 | ||||||||||||||||||||
Literature Links: |
PLEKHG6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLEKHG6 - pleckstrin homology and RhoGEF domain containing G6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144856.1 | 1560 | Intron | NP_001138328.1 | |||
NM_001144857.1 | 1560 | Intron | NP_001138329.1 | |||
NM_018173.3 | 1560 | Intron | NP_060643.2 | |||
XM_005253704.4 | 1560 | Intron | XP_005253761.1 | |||
XM_006718985.3 | 1560 | Intron | XP_006719048.1 | |||
XM_011520967.2 | 1560 | Intron | XP_011519269.1 | |||
XM_017019555.1 | 1560 | Intron | XP_016875044.1 |
TNFRSF1A - TNF receptor superfamily member 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001065.3 | 1560 | Silent Mutation | GAA,GAG | E,E 419 | NP_001056.1 |