Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGAAAGGTAACTCTGGGAAGTAC[C/T]CATATTCGGTAACTTACAGCTCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610148 MIM: 606736 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BBS10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BBS10 - Bardet-Biedl syndrome 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024685.3 | 1834 | Missense Mutation | AGT,GGT | S,G 584 | NP_078961.3 |
OSBPL8 - oxysterol binding protein like 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |