Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCGTGGGGTGCCGAGGCCCGGC[A/G]GCGAGGCGGTGGTGGAGGGGGCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614239 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM109A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM109A - family with sequence similarity 109 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177996.1 | 1019 | Missense Mutation | CGC,TGC | R,C 223 | NP_001171467.1 | |
NM_001177997.1 | 1019 | Missense Mutation | CGC,TGC | R,C 210 | NP_001171468.1 | |
NM_144671.4 | 1019 | Missense Mutation | CGC,TGC | R,C 210 | NP_653272.2 | |
XM_006719257.3 | 1019 | Missense Mutation | CGC,TGC | R,C 210 | XP_006719320.1 | |
XM_011537976.2 | 1019 | Missense Mutation | CGC,TGC | R,C 210 | XP_011536278.1 | |
XM_017018883.1 | 1019 | Missense Mutation | CGC,TGC | R,C 210 | XP_016874372.1 |