Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGGAGAGATCCACAACTGGACC[A/G]AGCTGCTTGACCTCTTCAACCACAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605307 | ||||||||||||||||||||
Literature Links: |
GPR182 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR182 - G protein-coupled receptor 182 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007264.3 | 861 | Missense Mutation | AAG,GAG | K,E 31 | NP_009195.1 | |
XM_011537824.2 | 861 | Missense Mutation | AAG,GAG | K,E 31 | XP_011536126.1 | |
XM_011537825.2 | 861 | Missense Mutation | AAG,GAG | K,E 31 | XP_011536127.1 | |
XM_011537827.2 | 861 | Missense Mutation | AAG,GAG | K,E 31 | XP_011536129.1 |
HBCBP - HBcAg-binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB39 - zinc finger and BTB domain containing 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |