Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCGTCTTAGCTTCTTCAGATCTA[G/T]ATCGACATCCTCAGGAACTTCGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608364 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIMA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIMA1 - LIM domain and actin binding 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113546.1 | 1775 | Silent Mutation | NP_001107018.1 | |||
NM_001113547.1 | 1775 | Silent Mutation | NP_001107019.1 | |||
NM_001243775.1 | 1775 | Silent Mutation | NP_001230704.1 | |||
NM_016357.4 | 1775 | Silent Mutation | NP_057441.1 | |||
XM_011538455.2 | 1775 | Missense Mutation | XP_011536757.1 | |||
XM_017019429.1 | 1775 | Silent Mutation | XP_016874918.1 | |||
XM_017019430.1 | 1775 | Silent Mutation | XP_016874919.1 |