Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTTGCAGACTTGGGACCCAGCA[C/G]TAGCCCAAATTGCAAAAGCATGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602692 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLIPR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLIPR1 - GLI pathogenesis related 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006851.2 | 317 | Silent Mutation | CTA,GTA | L,V 64 | NP_006842.2 | |
XM_011537775.2 | 317 | Missense Mutation | CTA,GTA | L,V 64 | XP_011536077.1 | |
XM_017018721.1 | 317 | UTR 5 | XP_016874210.1 |