Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCGGGCTTCACACAGTTCTGTCC[A/G]TCCCTGAGTGCTTCCTCACCACCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605884 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC92 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC92 - coiled-coil domain containing 92 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304957.1 | 1404 | Missense Mutation | ACG,ATG | T,M 327 | NP_001291886.1 | |
NM_001304958.1 | 1404 | Missense Mutation | ACG,ATG | T,M 327 | NP_001291887.1 | |
NM_001304959.1 | 1404 | Missense Mutation | ACG,ATG | T,M 327 | NP_001291888.1 | |
NM_001304960.1 | 1404 | Missense Mutation | ACG,ATG | T,M 327 | NP_001291889.1 | |
NM_001304961.1 | 1404 | Missense Mutation | ACG,ATG | T,M 310 | NP_001291890.1 | |
NM_025140.2 | 1404 | Missense Mutation | ACG,ATG | T,M 327 | NP_079416.1 | |
XM_005253624.2 | 1404 | Missense Mutation | ACG,ATG | T,M 327 | XP_005253681.1 | |
XM_017019984.1 | 1404 | Missense Mutation | ACG,ATG | T,M 327 | XP_016875473.1 |
DNAH10 - dynein axonemal heavy chain 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |