Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCTTCTTTGAGTTTTCCAATCA[C/T]TTCCTCTTTATCTTGCATCAGTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601936 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAWR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAWR - pro-apoptotic WT1 regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002583.2 | 1061 | Missense Mutation | ATG,GTG | M,V 299 | NP_002574.2 | |
XM_006719435.2 | 1061 | Missense Mutation | ATG,GTG | M,V 299 | XP_006719498.1 | |
XM_006719436.3 | 1061 | Intron | XP_006719499.1 | |||
XM_017019377.1 | 1061 | Missense Mutation | ATG,GTG | M,V 299 | XP_016874866.1 | |
XM_017019378.1 | 1061 | Intron | XP_016874867.1 | |||
XM_017019379.1 | 1061 | Intron | XP_016874868.1 |