Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGTTGACCAGATTAATCCATCT[A/G]TTTTCTGCATTCATATTACAAACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609863 | ||||||||||||||||||||
Literature Links: |
TCTN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TCTN1 - tectonic family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001082537.2 | 604 | Missense Mutation | ATT,GTT | I,V 150 | NP_001076006.1 | |
NM_001082538.2 | 604 | Missense Mutation | ATT,GTT | I,V 150 | NP_001076007.1 | |
NM_001173975.2 | 604 | Missense Mutation | ATT,GTT | I,V 94 | NP_001167446.1 | |
NM_001173976.1 | 604 | Missense Mutation | ATT,GTT | I,V 90 | NP_001167447.1 | |
NM_001319680.1 | 604 | Missense Mutation | ATT,GTT | I,V 150 | NP_001306609.1 | |
NM_001319681.1 | 604 | UTR 5 | NP_001306610.1 | |||
NM_001319682.1 | 604 | Missense Mutation | ATT,GTT | I,V 94 | NP_001306611.1 | |
NM_024549.5 | 604 | Missense Mutation | ATT,GTT | I,V 150 | NP_078825.2 | |
XM_005253934.3 | 604 | Missense Mutation | ATT,GTT | I,V 150 | XP_005253991.1 | |
XM_005253935.3 | 604 | Missense Mutation | ATT,GTT | I,V 150 | XP_005253992.1 | |
XM_006719594.2 | 604 | Missense Mutation | ATT,GTT | I,V 94 | XP_006719657.1 | |
XM_006719595.2 | 604 | UTR 5 | XP_006719658.1 | |||
XM_006719596.2 | 604 | UTR 5 | XP_006719659.1 | |||
XM_006719597.3 | 604 | UTR 5 | XP_006719660.1 | |||
XM_006719598.2 | 604 | UTR 5 | XP_006719661.1 | |||
XM_006719599.2 | 604 | UTR 5 | XP_006719662.1 | |||
XM_006719600.2 | 604 | UTR 5 | XP_006719663.1 | |||
XM_011538733.2 | 604 | Missense Mutation | ATT,GTT | I,V 150 | XP_011537035.1 | |
XM_011538734.2 | 604 | Missense Mutation | ATT,GTT | I,V 130 | XP_011537036.1 | |
XM_011538735.1 | 604 | Missense Mutation | ATT,GTT | I,V 150 | XP_011537037.1 | |
XM_011538737.2 | 604 | Missense Mutation | ATT,GTT | I,V 150 | XP_011537039.1 | |
XM_011538738.2 | 604 | Missense Mutation | ATT,GTT | I,V 150 | XP_011537040.1 | |
XM_017019964.1 | 604 | Missense Mutation | ATT,GTT | I,V 94 | XP_016875453.1 | |
XM_017019965.1 | 604 | UTR 5 | XP_016875454.1 | |||
XM_017019966.1 | 604 | UTR 5 | XP_016875455.1 | |||
XM_017019967.1 | 604 | UTR 5 | XP_016875456.1 | |||
XM_017019968.1 | 604 | UTR 5 | XP_016875457.1 | |||
XM_017019969.1 | 604 | UTR 5 | XP_016875458.1 |