Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCTTTCCCGTTTGAGGTATGGCA[C/T]TTTTCATTATCCTAGCCACGTTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 189904 | ||||||||||||||||||||
Literature Links: |
NFYB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NFYB - nuclear transcription factor Y subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006166.3 | 1129 | Missense Mutation | NP_006157.1 | |||
XM_005268908.4 | 1129 | Missense Mutation | XP_005268965.1 | |||
XM_005268909.4 | 1129 | Missense Mutation | XP_005268966.1 | |||
XM_005268910.4 | 1129 | Missense Mutation | XP_005268967.1 |