Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCATCACAGTTCTGGTTGATTTC[A/G]TGAAGCCACCGCTTGACGTTGACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 617002 MIM: 604199 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BICDL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BICDL1 - BICD family like cargo adaptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB35 - RAB35, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167606.1 | 403 | Silent Mutation | CAC,CAT | H,H 104 | NP_001161078.1 | |
NM_006861.6 | 403 | Silent Mutation | CAC,CAT | H,H 104 | NP_006852.1 | |
XM_011537779.1 | 403 | UTR 5 | XP_011536081.1 | |||
XM_017018723.1 | 403 | UTR 5 | XP_016874212.1 |