Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTGATGCACTGCAAGCTCAAGC[A/G]CTTTAATTCTAAAGCTTATGCTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602892 MIM: 602893 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLRC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLRC3 - killer cell lectin like receptor C3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002261.2 | 749 | Intron | NP_002252.2 | |||
NM_007333.2 | 749 | Missense Mutation | GCG,GTG | A,V 235 | NP_031359.2 |
KLRC4 - killer cell lectin like receptor C4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLRC4-KLRK1 - KLRC4-KLRK1 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |