Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTGACATGCGTGGTGGAGCTGC[A/G]TCGCCTCGTCTTGCCCTGCACCATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612664 | ||||||||||||||||||||
Literature Links: |
RERG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RERG - RAS like estrogen regulated growth inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190726.1 | 878 | Missense Mutation | CGC,TGC | R,C 161 | NP_001177655.1 | |
NM_032918.2 | 878 | Missense Mutation | CGC,TGC | R,C 180 | NP_116307.1 | |
XM_017020121.1 | 878 | Missense Mutation | CGC,TGC | R,C 177 | XP_016875610.1 |