Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCACCCACCTTCAGGAAGTCCTC[A/G]AAGGTGATCCCCTCGTACACCTGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609073 MIM: 611585 | ||||||||||||||||||||
Literature Links: |
FBXW8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXW8 - F-box and WD repeat domain containing 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TESC - tescalcin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168325.1 | 716 | Silent Mutation | TTC,TTT | F,F 157 | NP_001161797.1 | |
NM_017899.3 | 716 | Silent Mutation | TTC,TTT | F,F 184 | NP_060369.3 | |
XM_011538503.1 | 716 | Silent Mutation | TTC,TTT | F,F 172 | XP_011536805.1 | |
XM_017019532.1 | 716 | Intron | XP_016875021.1 | |||
XM_017019533.1 | 716 | Intron | XP_016875022.1 |