Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCACCATGGTGACGCTTGCTGA[A/G]CTGCTGGTGCTTCTGGCCGCTCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927415 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101927415 - uncharacterized LOC101927415 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMED2 - transmembrane p24 trafficking protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321445.1 | 126 | Silent Mutation | GAA,GAG | E,E 6 | NP_001308374.1 | |
NM_006815.3 | 126 | Silent Mutation | GAA,GAG | E,E 6 | NP_006806.1 |