Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACGTGCTGCCTGAGCCGGAGACGG[C/G]GAGCCGCTGCAACCCCGCGGCATCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604259 | ||||||||||||||||||||
Literature Links: |
PLXNC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLXNC1 - plexin C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005761.2 | 848 | Missense Mutation | GCG,GGG | A,G 191 | NP_005752.1 | |
XM_006719186.3 | 848 | Missense Mutation | GCG,GGG | A,G 191 | XP_006719249.1 | |
XM_011537730.2 | 848 | Missense Mutation | GCG,GGG | A,G 191 | XP_011536032.1 | |
XM_011537731.2 | 848 | Missense Mutation | GCG,GGG | A,G 191 | XP_011536033.1 | |
XM_017018671.1 | 848 | Missense Mutation | GCG,GGG | A,G 191 | XP_016874160.1 | |
XM_017018672.1 | 848 | Missense Mutation | GCG,GGG | A,G 191 | XP_016874161.1 |