Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCCCCCAGGGCCCGGACTCTCGC[G/T]GCTGCCGCTGCCGCTGCTGCTGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602285 | ||||||||||||||||||||
Literature Links: |
MMP17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MMP17 - matrix metallopeptidase 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016155.5 | 161 | Missense Mutation | CGG,CTG | R,L 20 | NP_057239.4 | |
XM_011538355.2 | 161 | Intron | XP_011536657.1 | |||
XM_011538356.2 | 161 | Intron | XP_011536658.1 | |||
XM_011538357.2 | 161 | Intron | XP_011536659.1 | |||
XM_017019307.1 | 161 | Missense Mutation | CGG,CTG | R,L 20 | XP_016874796.1 |