Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAACAGGCTACTCCTTTGTAAAT[A/T]GCAAGAAAGCTCTGGAGACTTGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615258 MIM: 604723 | ||||||||||||||||||||
Literature Links: |
METTL21B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
METTL21B - methyltransferase like 21B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015433.2 | 247 | Intron | NP_056248.2 | |||
NM_206914.1 | 247 | Intron | NP_996797.1 |
TSFM - Ts translation elongation factor, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172695.1 | 247 | Missense Mutation | AGC,TGC | S,C 64 | NP_001166166.1 | |
NM_001172696.1 | 247 | Missense Mutation | AGC,TGC | S,C 64 | NP_001166167.1 | |
NM_001172697.1 | 247 | Missense Mutation | AGC,TGC | S,C 64 | NP_001166168.1 | |
NM_005726.5 | 247 | Missense Mutation | AGC,TGC | S,C 64 | NP_005717.3 |