Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGGTTCCTTACTGCCTCTCTC[A/G]TGAGTTTTAGGGATGCATTCTGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 171891 MIM: 613246 | ||||||||||||||||||||
Literature Links: |
PDE1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDE1B - phosphodiesterase 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R1A - protein phosphatase 1 regulatory inhibitor subunit 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006741.3 | 427 | Silent Mutation | NP_006732.3 | |||
XM_005268995.4 | 427 | Silent Mutation | XP_005269052.1 | |||
XM_006719471.3 | 427 | Missense Mutation | XP_006719534.1 |