Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTTCTCGGGGCAAGAGGACTCAC[C/T]GGTTTTGCACCTGGACTGCACAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611161 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT80 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRT80 - keratin 80 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081492.1 | 1332 | Missense Mutation | ACC,GCC | T,A 412 | NP_001074961.1 | |
NM_182507.2 | 1332 | Missense Mutation | ACT,GCT | T,A 412 | NP_872313.2 | |
XM_005268676.3 | 1332 | Missense Mutation | ACT,GCT | T,A 447 | XP_005268733.1 |