Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTCCACCGAGAGCCGGTAAGCC[A/G]GGACCAGCCCCAGCCCCACTGCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147559 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGB7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITGB7 - integrin subunit beta 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000889.2 | 2463 | Missense Mutation | CCG,CTG | P,L 744 | NP_000880.1 | |
XM_005268851.3 | 2463 | Missense Mutation | CCG,CTG | P,L 744 | XP_005268908.1 | |
XM_005268852.4 | 2463 | Missense Mutation | CCG,CTG | P,L 744 | XP_005268909.1 | |
XM_006719376.3 | 2463 | Missense Mutation | CCG,CTG | P,L 744 | XP_006719439.1 |
ZNF740 - zinc finger protein 740 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |