Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGGGCAGCGGGGCCGGGCTTAC[C/T]GCGCTTCAGCCTCTCCATGGCACTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613541 MIM: 602198 | ||||||||||||||||||||
Literature Links: |
C12orf65 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C12orf65 - chromosome 12 open reading frame 65 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CDK2AP1 - cyclin dependent kinase 2 associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270433.1 | 802 | Intron | NP_001257362.1 | |||
NM_001270434.1 | 802 | Intron | NP_001257363.1 | |||
NM_004642.3 | 802 | Missense Mutation | AGC,GGC | S,G 94 | NP_004633.1 |