Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTACAGACTTATGGCCTTCACCAC[C/T]GGGAGGAGCAGCGTGTGGAAATGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612027 | ||||||||||||||||||||
Literature Links: |
GRASP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRASP - general receptor for phosphoinositides 1 associated scaffold protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271856.1 | 145 | UTR 5 | NP_001258785.1 | |||
NM_181711.3 | 145 | Missense Mutation | CGG,TGG | R,W 124 | NP_859062.1 | |
XM_005268691.3 | 145 | UTR 5 | XP_005268748.1 | |||
XM_011537996.1 | 145 | UTR 5 | XP_011536298.1 |