Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACACCCTTAAAATCTATTTACC[A/G]CTTGGAAGTTAGAAGTAAGGTAATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616381 | ||||||||||||||||||||
Literature Links: |
ZCCHC8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZCCHC8 - zinc finger CCHC-type containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017612.4 | 1379 | Missense Mutation | NP_060082.2 | |||
XM_006719491.2 | 1379 | Missense Mutation | XP_006719554.1 | |||
XM_011538555.2 | 1379 | Missense Mutation | XP_011536857.1 | |||
XM_017019606.1 | 1379 | Missense Mutation | XP_016875095.1 |