Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTACCTAAGGAACACCAATCTAG[C/G]CCCTAGCCTTCAGCTATGTCAGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604143 | ||||||||||||||||||||
Literature Links: |
ESPL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ESPL1 - extra spindle pole bodies like 1, separase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012291.4 | 954 | Missense Mutation | GCC,GGC | A,G 288 | NP_036423.4 | |
XM_006719705.3 | 954 | Missense Mutation | GCC,GGC | A,G 288 | XP_006719768.1 | |
XM_011539024.2 | 954 | Missense Mutation | GCC,GGC | A,G 288 | XP_011537326.1 | |
XM_011539025.2 | 954 | Missense Mutation | GCC,GGC | A,G 288 | XP_011537327.1 | |
XM_017020253.1 | 954 | UTR 5 | XP_016875742.1 |