Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGGCAGTGTCAAAACGACATCTA[C/T]TTCTTTGCTTGCTTTGTGATCATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176914 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1CC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1CC - protein phosphatase 1 catalytic subunit gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244974.1 | 1240 | Intron | NP_001231903.1 | |||
NM_002710.3 | 1240 | Silent Mutation | AAA,AAG | K,K 323 | NP_002701.1 | |
XM_011538504.2 | 1240 | Intron | XP_011536806.1 | |||
XM_011538505.2 | 1240 | Intron | XP_011536807.1 |