Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGGCTACACGGACTCAGCCCCA[C/G]AGCGGCCTCCTCATCCTCTGTGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CALCOCO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CALCOCO1 - calcium binding and coiled-coil domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143682.1 | 1811 | Missense Mutation | CTG,GTG | L,V 443 | NP_001137154.1 | |
NM_020898.2 | 1811 | Missense Mutation | CTG,GTG | L,V 528 | NP_065949.1 | |
XM_011538601.1 | 1811 | Missense Mutation | CTG,GTG | L,V 528 | XP_011536903.1 | |
XM_011538602.1 | 1811 | Missense Mutation | CTG,GTG | L,V 528 | XP_011536904.1 | |
XM_011538603.1 | 1811 | Missense Mutation | CTG,GTG | L,V 528 | XP_011536905.1 | |
XM_011538604.1 | 1811 | Missense Mutation | CTG,GTG | L,V 528 | XP_011536906.1 | |
XM_017019707.1 | 1811 | Missense Mutation | CTG,GTG | L,V 528 | XP_016875196.1 | |
XM_017019708.1 | 1811 | Missense Mutation | CTG,GTG | L,V 205 | XP_016875197.1 | |
XM_017019709.1 | 1811 | Missense Mutation | CTG,GTG | L,V 205 | XP_016875198.1 |