Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACTTGCCAAAGTTAAACAAACT[C/T]AAGAAGCTTGAACTAAGCAGTAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606878 | ||||||||||||||||||||
Literature Links: |
ANP32D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANP32D - acidic nuclear phosphoprotein 32 family member D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012404.2 | Intron | NP_036536.2 |
C12orf54 - chromosome 12 open reading frame 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152319.3 | Intron | NP_689532.1 | ||||
XM_005268636.3 | Intron | XP_005268693.1 | ||||
XM_011537896.2 | Intron | XP_011536198.1 | ||||
XM_017018796.1 | Intron | XP_016874285.1 |