Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCAAAGTTATGAATGCAGATTGC[C/T]TCCACAAATTCACTGCTGGTACCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616706 | ||||||||||||||||||||
Literature Links: |
PARP11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PARP11 - poly(ADP-ribose) polymerase family member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286521.1 | 684 | Silent Mutation | NP_001273450.1 | |||
NM_001286522.1 | 684 | Silent Mutation | NP_001273451.1 | |||
NM_020367.5 | 684 | Silent Mutation | NP_065100.2 | |||
XM_005253711.4 | 684 | Silent Mutation | XP_005253768.1 | |||
XM_011520970.1 | 684 | Silent Mutation | XP_011519272.1 | |||
XM_011520971.2 | 684 | Missense Mutation | XP_011519273.1 | |||
XM_011520973.2 | 684 | Silent Mutation | XP_011519275.1 | |||
XM_017019670.1 | 684 | Silent Mutation | XP_016875159.1 | |||
XM_017019671.1 | 684 | Silent Mutation | XP_016875160.1 |