Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCAGGTTTACTTCGGGGGCCGGGA[A/G]CCCCAAGATGCGGAAAGTGCGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610588 MIM: 602742 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDN - dendrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015086.1 | 2274 | Missense Mutation | CTC,TTC | L,F 551 | NP_055901.2 | |
XM_011538055.2 | 2274 | Missense Mutation | CTC,TTC | L,F 497 | XP_011536357.1 |
PRKAG1 - protein kinase AMP-activated non-catalytic subunit gamma 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |