Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTTTTTTCTTTTTCAGTGATG[A/G]TCATGTGAAGTTTTTCTGCTTTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603180 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
XPOT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
XPOT - exportin for tRNA | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007235.4 | 655 | Missense Mutation | GAT,GGT | D,G 50 | NP_009166.2 | |
XM_017018748.1 | 655 | Missense Mutation | GAT,GGT | D,G 50 | XP_016874237.1 |