Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGATTCAATTCCTGGAGAGAAAC[A/C]CCTGCCACATTCACACCACTCCACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 150100 | ||||||||||||||||||||
Literature Links: |
LDHB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LDHB - lactate dehydrogenase B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174097.2 | 854 | Silent Mutation | GGG,GGT | G,G 209 | NP_001167568.1 | |
NM_001315537.1 | 854 | Intron | NP_001302466.1 | |||
NM_002300.7 | 854 | Intron | NP_002291.1 | |||
XM_006719074.2 | 854 | Silent Mutation | GGG,GGT | G,G 209 | XP_006719137.1 |