Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCAGGCAGCGGCCCGTGGCCCG[A/G]CTCACAATGGGGCCACTCTGTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606251 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALNT9 - polypeptide N-acetylgalactosaminyltransferase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122636.1 | 863 | Silent Mutation | AGC,AGT | S,S 561 | NP_001116108.1 | |
NM_021808.3 | 863 | Silent Mutation | AGC,AGT | S,S 195 | NP_068580.2 | |
XM_017019371.1 | 863 | Silent Mutation | AGC,AGT | S,S 460 | XP_016874860.1 | |
XM_017019372.1 | 863 | Silent Mutation | AGC,AGT | S,S 458 | XP_016874861.1 | |
XM_017019373.1 | 863 | Silent Mutation | AGC,AGT | S,S 458 | XP_016874862.1 | |
XM_017019374.1 | 863 | Silent Mutation | AGC,AGT | S,S 418 | XP_016874863.1 | |
XM_017019375.1 | 863 | Intron | XP_016874864.1 | |||
XM_017019376.1 | 863 | Intron | XP_016874865.1 |