Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGAGTTTTCAGTTCCAGGGCCCA[A/G]ATCCTGAGGAGTCTGGATGGGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600407 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RFC5 - replication factor C subunit 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WSB2 - WD repeat and SOCS box containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278557.1 | 1020 | Silent Mutation | ATC,ATT | I,I 336 | NP_001265486.1 | |
NM_001278558.1 | 1020 | Silent Mutation | ATC,ATT | I,I 109 | NP_001265487.1 | |
NM_018639.4 | 1020 | Silent Mutation | ATC,ATT | I,I 319 | NP_061109.1 | |
XM_017019642.1 | 1020 | Silent Mutation | ATC,ATT | I,I 109 | XP_016875131.1 |