Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGGGTGAGCAGTCAGTATTCAAA[C/T]GTGGTTTCTGTGGCTGAGAGAGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606092 MIM: 601807 | ||||||||||||||||||||
Literature Links: |
DNAJC14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJC14 - DnaJ heat shock protein family (Hsp40) member C14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMP19 - matrix metallopeptidase 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272101.1 | 1408 | UTR 3 | NP_001259030.1 | |||
NM_002429.5 | 1408 | Silent Mutation | ACA,ACG | T,T 505 | NP_002420.1 | |
XM_006719401.3 | 1408 | Silent Mutation | ACA,ACG | T,T 276 | XP_006719464.1 | |
XM_011538359.1 | 1408 | Silent Mutation | ACA,ACG | T,T 423 | XP_011536661.1 | |
XM_017019308.1 | 1408 | Intron | XP_016874797.1 | |||
XM_017019309.1 | 1408 | Intron | XP_016874798.1 |
TMEM198B - transmembrane protein 198B (pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |