Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGGTGGGCGGGGCTTACATGCC[C/G]GGCACCACCCCATTGGTCTCCATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609695 MIM: 611055 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HPD - 4-hydroxyphenylpyruvate dioxygenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171993.1 | 1213 | Silent Mutation | CCC,CCG | P,P 352 | NP_001165464.1 | |
NM_002150.2 | 1213 | Silent Mutation | CCC,CCG | P,P 391 | NP_002141.1 |
SETD1B - SET domain containing 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |