Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACCTTAGGGATAATATGCCTTTC[A/G]TTGATGTCTACGTTGGGAATTTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602894 | ||||||||||||||||||||
Literature Links: |
KLRD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLRD1 - killer cell lectin like receptor D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114396.1 | 2124 | Silent Mutation | NP_001107868.1 | |||
NM_002262.3 | 2124 | Silent Mutation | TCA,TCG | S,S 22 | NP_002253.1 | |
NM_007334.2 | 2124 | Intron | NP_031360.1 | |||
XM_006719067.3 | 2124 | Silent Mutation | TCA,TCG | S,S 22 | XP_006719130.1 | |
XM_011520650.2 | 2124 | Intron | XP_011518952.1 | |||
XM_011520651.2 | 2124 | Silent Mutation | TCA,TCG | S,S 22 | XP_011518953.1 | |
XM_017019285.1 | 2124 | Silent Mutation | TCA,TCG | S,S 22 | XP_016874774.1 | |
XM_017019286.1 | 2124 | Silent Mutation | TCA,TCG | S,S 22 | XP_016874775.1 | |
XM_017019287.1 | 2124 | Silent Mutation | TCA,TCG | S,S 22 | XP_016874776.1 | |
XM_017019288.1 | 2124 | Intron | XP_016874777.1 | |||
XM_017019289.1 | 2124 | Intron | XP_016874778.1 | |||
XM_017019290.1 | 2124 | Silent Mutation | TCA,TCG | S,S 22 | XP_016874779.1 | |
XM_017019291.1 | 2124 | Silent Mutation | TCA,TCG | S,S 22 | XP_016874780.1 |