Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTCACCCCTGCGGAGGCGGCCCA[C/T]GGTATGCGAGAAGCCATAATACTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605427 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPV4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRPV4 - transient receptor potential cation channel subfamily V member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177428.1 | 2338 | Missense Mutation | NP_001170899.1 | |||
NM_001177431.1 | 2338 | Missense Mutation | NP_001170902.1 | |||
NM_001177433.1 | 2338 | Missense Mutation | NP_001170904.1 | |||
NM_021625.4 | 2338 | Missense Mutation | NP_067638.3 | |||
NM_147204.2 | 2338 | Missense Mutation | NP_671737.1 | |||
XM_005253918.1 | 2338 | Missense Mutation | XP_005253975.1 | |||
XM_011538630.2 | 2338 | Missense Mutation | XP_011536932.2 | |||
XM_011538631.2 | 2338 | Missense Mutation | XP_011536933.2 | |||
XM_011538632.2 | 2338 | Missense Mutation | XP_011536934.2 | |||
XM_011538633.2 | 2338 | Missense Mutation | XP_011536935.2 | |||
XM_011538634.2 | 2338 | Intron | XP_011536936.2 | |||
XM_011538635.2 | 2338 | Intron | XP_011536937.1 | |||
XM_011538636.2 | 2338 | Intron | XP_011536938.1 | |||
XM_017019774.1 | 2338 | Missense Mutation | XP_016875263.1 |