Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAATAACCAGGTCCAGCTTATTCC[C/T]GGATTCTGCTATGAGGGGCACAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604597 MIM: 614539 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRIP1 - glutamate receptor interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178074.1 | 3384 | Missense Mutation | AGG,GGG | R,G 1008 | NP_001171545.1 | |
NM_021150.3 | 3384 | Missense Mutation | AGG,GGG | R,G 1023 | NP_066973.2 | |
XM_005268754.4 | 3384 | Missense Mutation | AGG,GGG | R,G 1076 | XP_005268811.1 | |
XM_005268757.4 | 3384 | Missense Mutation | AGG,GGG | R,G 1049 | XP_005268814.1 | |
XM_011538093.2 | 3384 | Missense Mutation | AGG,GGG | R,G 1075 | XP_011536395.1 | |
XM_017019098.1 | 3384 | Missense Mutation | AGG,GGG | R,G 1135 | XP_016874587.1 | |
XM_017019099.1 | 3384 | Missense Mutation | AGG,GGG | R,G 1101 | XP_016874588.1 | |
XM_017019100.1 | 3384 | Missense Mutation | AGG,GGG | R,G 1083 | XP_016874589.1 | |
XM_017019101.1 | 3384 | Missense Mutation | AGG,GGG | R,G 1049 | XP_016874590.1 | |
XM_017019102.1 | 3384 | Missense Mutation | AGG,GGG | R,G 1048 | XP_016874591.1 |
HELB - DNA helicase B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |