Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAGGCCCTAAAGCTTTTTAAGA[A/C]CCAGCTGTCTCCATATGAACAAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609181 | ||||||||||||||||||||
Literature Links: |
DYRK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DYRK4 - dual specificity tyrosine phosphorylation regulated kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282285.1 | 306 | Intron | NP_001269214.1 | |||
NM_001282286.1 | 306 | Intron | NP_001269215.1 | |||
NM_003845.2 | 306 | Missense Mutation | AAC,ACC | N,T 47 | NP_003836.1 |