Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTGTTTTAAACTATTTGTAGGC[G/T]CAGTCATGCAGGAAAACCTCAGATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600372 | ||||||||||||||||||||
Literature Links: |
NEDD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NEDD1 - neural precursor cell expressed, developmentally down-regulated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135175.1 | 334 | Missense Mutation | GCA,TCA | A,S 6 | NP_001128647.1 | |
NM_001135176.1 | 334 | UTR 5 | NP_001128648.1 | |||
NM_001135177.1 | 334 | Intron | NP_001128649.1 | |||
NM_152905.3 | 334 | UTR 5 | NP_690869.1 | |||
XM_005268644.2 | 334 | Missense Mutation | GCA,TCA | A,S 6 | XP_005268701.1 | |
XM_006719237.3 | 334 | Intron | XP_006719300.1 | |||
XM_011537903.2 | 334 | Intron | XP_011536205.1 | |||
XM_011537904.2 | 334 | UTR 5 | XP_011536206.1 | |||
XM_011537905.2 | 334 | Intron | XP_011536207.1 | |||
XM_011537906.2 | 334 | Intron | XP_011536208.1 | |||
XM_017018801.1 | 334 | Intron | XP_016874290.1 | |||
XM_017018802.1 | 334 | Intron | XP_016874291.1 |