Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTTTTTTCTTTCTTCTTTGGAG[G/T]CTCACGTTCTCCGAGCATATTTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610750 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR7851 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR7851 - microRNA 7851 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZCRB1 - zinc finger CCHC-type and RNA binding motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033114.3 | 488 | Missense Mutation | NP_149105.3 | |||
XM_017020124.1 | 488 | Missense Mutation | XP_016875613.1 |