Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCGGGCTGGCGGCCAGGGCGGC[A/G]ACTTTGCCGTCCAGGGGCCCCGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601621 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBX3 - T-box 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005996.3 | 2926 | Silent Mutation | GTC,GTT | V,V 654 | NP_005987.3 | |
NM_016569.3 | 2926 | Silent Mutation | GTC,GTT | V,V 674 | NP_057653.3 |