Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATCAAGAGAATGATGATGAGGG[A/G]GAAGAAGATAAATTGTTTTGGCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609007 | ||||||||||||||||||||
Literature Links: |
LRRK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRRK2 - leucine rich repeat kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198578.3 | 1263 | Silent Mutation | GGA,GGG | G,G 342 | NP_940980.3 | |
XM_005268629.3 | 1263 | Silent Mutation | GGA,GGG | G,G 342 | XP_005268686.1 | |
XM_011537877.2 | 1263 | Silent Mutation | GGA,GGG | G,G 342 | XP_011536179.1 | |
XM_011537881.2 | 1263 | Silent Mutation | GGA,GGG | G,G 342 | XP_011536183.1 | |
XM_011537882.2 | 1263 | Silent Mutation | GGA,GGG | G,G 342 | XP_011536184.1 | |
XM_017018786.1 | 1263 | Silent Mutation | GGA,GGG | G,G 342 | XP_016874275.1 | |
XM_017018787.1 | 1263 | Intron | XP_016874276.1 | |||
XM_017018788.1 | 1263 | Intron | XP_016874277.1 | |||
XM_017018789.1 | 1263 | Silent Mutation | GGA,GGG | G,G 342 | XP_016874278.1 |