Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCGTGCGGAAGGGGAGGCTTGAG[A/G]GGGACCAGGAAGGGTGGAAGTCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSRNP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSRNP2 - cysteine and serine rich nuclear protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030809.2 | 1741 | Missense Mutation | CCC,CTC | P,L 505 | NP_110436.1 | |
XM_006719621.2 | 1741 | Missense Mutation | CCC,CTC | P,L 505 | XP_006719684.1 | |
XM_017019991.1 | 1741 | Missense Mutation | CCC,CTC | P,L 266 | XP_016875480.1 |