Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTAGAGTCAGTGCGTCCTCATCC[A/G]CAGCTCCAGATGCTGTTCTGGTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616381 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZCCHC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZCCHC8 - zinc finger CCHC-type containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017612.4 | 1682 | Missense Mutation | GCG,GTG | A,V 510 | NP_060082.2 | |
XM_006719491.2 | 1682 | Missense Mutation | GCG,GTG | A,V 433 | XP_006719554.1 | |
XM_011538555.2 | 1682 | Missense Mutation | GCG,GTG | A,V 411 | XP_011536857.1 | |
XM_017019606.1 | 1682 | Missense Mutation | GCG,GTG | A,V 419 | XP_016875095.1 |